Product Details

SNP ID
rs147024648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23106130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCGGGTTTCATGGGCGTCATGA[A/G]TAACATGCGGAAACAGGTACCGCCT
Phenotype
MIM: 611119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL7 PubMed Links

Gene Details

Gene
KLHL7
Gene Name
kelch like family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031710.2 344 Missense Mutation AAT,AGT N35S NP_001026880.2
NM_001172428.1 344 Missense Mutation AAT,AGT N35S NP_001165899.1
NM_018846.4 344 Intron NP_061334.4
XM_006715753.2 344 Missense Mutation AAT,AGT N35S XP_006715816.1
XM_006715754.2 344 UTR 5 XP_006715817.1
XM_006715755.2 344 Intron XP_006715818.1
XM_006715756.2 344 Intron XP_006715819.1
XM_006715757.3 344 Missense Mutation AAT,AGT N35S XP_006715820.1
XM_017012439.1 344 Intron XP_016867928.1
XM_017012440.1 344 Missense Mutation AAT,AGT N35S XP_016867929.1
XM_017012441.1 344 UTR 5 XP_016867930.1
Gene
KLHL7-AS1
Gene Name
KLHL7 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details