Product Details

SNP ID
rs147169344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122702295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTTCAGGTTGCTTATCATCAC[C/T]GCGACTATATTTTCCGTCCCCTGGT
Phenotype
MIM: 609978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADPS2 PubMed Links

Gene Details

Gene
CADPS2
Gene Name
calcium dependent secretion activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009571.3 673 Intron NP_001009571.2
NM_001167940.1 673 Intron NP_001161412.1
NM_017954.10 673 Intron NP_060424.9
XM_005250695.4 673 Intron XP_005250752.1
XM_005250696.4 673 Intron XP_005250753.1
XM_005250697.4 673 Intron XP_005250754.1
XM_005250698.4 673 Intron XP_005250755.1
XM_005250699.4 673 Intron XP_005250756.1
XM_005250700.4 673 Intron XP_005250757.1
XM_005250701.4 673 Intron XP_005250758.1
XM_005250702.4 673 Intron XP_005250759.1
XM_005250703.4 673 Intron XP_005250760.1
XM_005250704.4 673 Intron XP_005250761.1
XM_005250705.4 673 Intron XP_005250762.1
XM_005250706.4 673 Intron XP_005250763.1
XM_005250707.4 673 Intron XP_005250764.1
XM_005250708.3 673 Intron XP_005250765.1
XM_011516695.2 673 Intron XP_011514997.1
XM_011516696.2 673 Intron XP_011514998.1
XM_011516697.2 673 Intron XP_011514999.1
XM_017012794.1 673 Intron XP_016868283.1
XM_017012795.1 673 Intron XP_016868284.1
XM_017012796.1 673 Intron XP_016868285.1
XM_017012797.1 673 Intron XP_016868286.1
XM_017012798.1 673 Intron XP_016868287.1
XM_017012799.1 673 Intron XP_016868288.1
XM_017012800.1 673 Intron XP_016868289.1
Gene
RNF133
Gene Name
ring finger protein 133
There are no transcripts associated with this gene.

Gene
RNF148
Gene Name
ring finger protein 148
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198085.1 673 Silent Mutation NP_932351.1

View Full Product Details