Product Details

SNP ID
rs147759063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93886826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACATATTAAGAATGTTTAAAA[C/T]TGCTTCTTCCGAATTTTCCGGATTC
Phenotype
MIM: 600033
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105375401 PubMed Links

Gene Details

Gene
LOC105375401
Gene Name
uncharacterized LOC105375401
There are no transcripts associated with this gene.

Gene
TFPI2
Gene Name
tissue factor pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271003.1 939 Silent Mutation CAA,CAG Q223Q NP_001257932.1
NM_001271004.1 939 UTR 3 NP_001257933.1
NM_006528.3 939 Silent Mutation CAA,CAG Q234Q NP_006519.1

View Full Product Details