Product Details

SNP ID
rs148007054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6161789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCTTAGGCTTGTTTTAACAAT[A/G]CATGTGATTTTTATTTAGTGAACAC
Phenotype
MIM: 605081
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYTH3 PubMed Links

Gene Details

Gene
CYTH3
Gene Name
cytohesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004227.3 4459 UTR 3 NP_004218.1
Gene
USP42
Gene Name
ubiquitin specific peptidase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032172.2 4459 Intron NP_115548.1
XM_005249883.4 4459 Intron XP_005249940.1
XM_006715790.1 4459 Intron XP_006715853.1
XM_006715791.2 4459 Intron XP_006715854.1
XM_011515573.1 4459 Intron XP_011513875.1
XM_011515574.1 4459 Intron XP_011513876.1
XM_011515577.1 4459 Intron XP_011513879.1
XM_011515578.1 4459 Intron XP_011513880.1

View Full Product Details