Product Details

SNP ID
rs148216190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88794339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGAGTAGGTCAGGTGCAACTT[C/G]GTGTAAGTTATCGCCTGGTCCTTTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C7orf62 PubMed Links

Gene Details

Gene
C7orf62
Gene Name
chromosome 7 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152706.3 786 Missense Mutation CAA,GAA Q202E NP_689919.1
XM_011515904.2 786 Missense Mutation CAA,GAA Q202E XP_011514206.1
Gene
ZNF804B
Gene Name
zinc finger protein 804B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181646.3 786 Intron NP_857597.1

View Full Product Details