Product Details
- SNP ID
-
rs148228353
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:148807676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATTTCCATTTCTCTTTCGATGCC[A/G]ACATACTTCAGGGCATCAGCCTGGC
- Phenotype
-
MIM: 603134
MIM: 601573
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CUL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs560966145] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CUL1
- Gene Name
- cullin 1
There are no transcripts associated with this gene.
- Gene
- EZH2
- Gene Name
- enhancer of zeste 2 polycomb repressive complex 2 subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001203247.1 |
4745 |
Silent Mutation |
GTC,GTT |
V737V |
NP_001190176.1 |
NM_001203248.1 |
4745 |
Silent Mutation |
GTC,GTT |
V728V |
NP_001190177.1 |
NM_001203249.1 |
4745 |
Silent Mutation |
GTC,GTT |
V686V |
NP_001190178.1 |
NM_004456.4 |
4745 |
Silent Mutation |
GTC,GTT |
V742V |
NP_004447.2 |
NM_152998.2 |
4745 |
Silent Mutation |
GTC,GTT |
V698V |
NP_694543.1 |
XM_005249962.4 |
4745 |
Silent Mutation |
GTC,GTT |
V745V |
XP_005250019.1 |
XM_005249963.4 |
4745 |
Silent Mutation |
GTC,GTT |
V736V |
XP_005250020.1 |
XM_005249964.4 |
4745 |
Silent Mutation |
GTC,GTT |
V694V |
XP_005250021.1 |
XM_011515883.2 |
4745 |
Silent Mutation |
GTC,GTT |
V750V |
XP_011514185.1 |
XM_011515884.2 |
4745 |
Silent Mutation |
GTC,GTT |
V742V |
XP_011514186.1 |
XM_011515885.2 |
4745 |
Silent Mutation |
GTC,GTT |
V741V |
XP_011514187.1 |
XM_011515886.2 |
4745 |
Silent Mutation |
GTC,GTT |
V734V |
XP_011514188.1 |
XM_011515887.2 |
4745 |
Silent Mutation |
GTC,GTT |
V733V |
XP_011514189.1 |
XM_011515888.2 |
4745 |
Silent Mutation |
GTC,GTT |
V733V |
XP_011514190.1 |
XM_011515889.2 |
4745 |
Silent Mutation |
GTC,GTT |
V720V |
XP_011514191.1 |
XM_011515890.2 |
4745 |
Silent Mutation |
GTC,GTT |
V711V |
XP_011514192.1 |
XM_011515891.2 |
4745 |
Silent Mutation |
GTC,GTT |
V709V |
XP_011514193.1 |
XM_011515892.2 |
4745 |
Silent Mutation |
GTC,GTT |
V708V |
XP_011514194.1 |
XM_011515893.2 |
4745 |
Silent Mutation |
GTC,GTT |
V706V |
XP_011514195.1 |
XM_011515894.2 |
4745 |
Silent Mutation |
GTC,GTT |
V703V |
XP_011514196.1 |
XM_011515895.2 |
4745 |
Silent Mutation |
GTC,GTT |
V702V |
XP_011514197.1 |
XM_011515896.2 |
4745 |
Silent Mutation |
GTC,GTT |
V664V |
XP_011514198.1 |
XM_011515897.2 |
4745 |
Silent Mutation |
GTC,GTT |
V633V |
XP_011514199.1 |
XM_011515898.2 |
4745 |
Silent Mutation |
GTC,GTT |
V633V |
XP_011514200.1 |
XM_011515899.2 |
4745 |
Intron |
|
|
XP_011514201.1 |
XM_011515901.2 |
4745 |
Intron |
|
|
XP_011514203.1 |
XM_017011817.1 |
4745 |
Silent Mutation |
GTC,GTT |
V750V |
XP_016867306.1 |
XM_017011818.1 |
4745 |
Silent Mutation |
GTC,GTT |
V729V |
XP_016867307.1 |
XM_017011819.1 |
4745 |
Silent Mutation |
GTC,GTT |
V703V |
XP_016867308.1 |
XM_017011820.1 |
4745 |
Silent Mutation |
GTC,GTT |
V694V |
XP_016867309.1 |
XM_017011821.1 |
4745 |
Silent Mutation |
GTC,GTT |
V628V |
XP_016867310.1 |
View Full Product Details