Product Details

SNP ID
rs148604231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:137386489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCTATAAGATGATCAAGGTCA[A/G]TGGACATCCATGTGTCCTTTGAGTA
Phenotype
MIM: 604072
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKI PubMed Links

Gene Details

Gene
DGKI
Gene Name
diacylglycerol kinase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321708.1 7655 Intron NP_001308637.1
NM_001321709.1 7655 Intron NP_001308638.1
NM_001321710.1 7655 Intron NP_001308639.1
NM_004717.3 7655 Intron NP_004708.1
XM_011516682.2 7655 Intron XP_011514984.2
XM_011516684.2 7655 UTR 3 XP_011514986.2
XM_017012780.1 7655 UTR 3 XP_016868269.1
XM_017012781.1 7655 UTR 3 XP_016868270.1
XM_017012782.1 7655 UTR 3 XP_016868271.1
XM_017012783.1 7655 UTR 3 XP_016868272.1
XM_017012784.1 7655 UTR 3 XP_016868273.1
XM_017012785.1 7655 UTR 3 XP_016868274.1
XM_017012786.1 7655 UTR 3 XP_016868275.1
XM_017012787.1 7655 UTR 3 XP_016868276.1
XM_017012788.1 7655 UTR 3 XP_016868277.1
XM_017012789.1 7655 UTR 3 XP_016868278.1

View Full Product Details