Product Details

SNP ID
rs148862996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144004258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTACTGTTTAGTTTTTGGTCT[A/G]TGAACAACAGCATCTCTTTCACACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6B1 PubMed Links

Gene Details

Gene
OR6B1
Gene Name
olfactory receptor family 6 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005281.1 262 Missense Mutation ATG,GTG M88V NP_001005281.1

View Full Product Details