Product Details

SNP ID
rs148896284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:12214912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAATGGACTGGTTAATAGTGA[A/G]GTCCATAATGAAGATGGAAGAAATG
Phenotype
MIM: 613413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM106B PubMed Links

Gene Details

Gene
TMEM106B
Gene Name
transmembrane protein 106B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134232.1 308 Silent Mutation GAA,GAG E34E NP_001127704.1
NM_018374.3 308 Silent Mutation GAA,GAG E34E NP_060844.2
XM_005249789.1 308 Silent Mutation GAA,GAG E34E XP_005249846.1

View Full Product Details