Product Details

SNP ID
rs149474376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128835534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTCAACCGTGACTGGCAGGA[C/T]GGCAAAGCTCTGGGCGCCCTGGTGG
Phenotype
MIM: 102565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLNC PubMed Links

Gene Details

Gene
FLNC
Gene Name
filamin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127487.1 770 Silent Mutation GAC,GAT D187D NP_001120959.1
NM_001458.4 770 Silent Mutation GAC,GAT D187D NP_001449.3

View Full Product Details