Product Details

SNP ID
hCV173438479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102399564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTACAGGTTCAAGTGCTGGGGC[C/T]GGCAGTGGAGAGTTCCACGTGTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100630923 PubMed Links

Gene Details

Gene
LOC100630923
Gene Name
LOC100289561-PRKRIP1 readthrough
There are no transcripts associated with this gene.

Gene
MIR548O
Gene Name
microRNA 548o
There are no transcripts associated with this gene.

Gene
PRKRIP1
Gene Name
PRKR interacting protein 1 (IL11 inducible)
There are no transcripts associated with this gene.

View Full Product Details