Product Details

SNP ID
rs150450224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122701909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAGGGGTCAATGCATGCCTTA[A/T]GGAAAAAATGTTTGCAAGTTAAAAT
Phenotype
MIM: 609978
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CADPS2 PubMed Links

Gene Details

Gene
CADPS2
Gene Name
calcium dependent secretion activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009571.3 1059 Intron NP_001009571.2
NM_001167940.1 1059 Intron NP_001161412.1
NM_017954.10 1059 Intron NP_060424.9
XM_005250695.4 1059 Intron XP_005250752.1
XM_005250696.4 1059 Intron XP_005250753.1
XM_005250697.4 1059 Intron XP_005250754.1
XM_005250698.4 1059 Intron XP_005250755.1
XM_005250699.4 1059 Intron XP_005250756.1
XM_005250700.4 1059 Intron XP_005250757.1
XM_005250701.4 1059 Intron XP_005250758.1
XM_005250702.4 1059 Intron XP_005250759.1
XM_005250703.4 1059 Intron XP_005250760.1
XM_005250704.4 1059 Intron XP_005250761.1
XM_005250705.4 1059 Intron XP_005250762.1
XM_005250706.4 1059 Intron XP_005250763.1
XM_005250707.4 1059 Intron XP_005250764.1
XM_005250708.3 1059 Intron XP_005250765.1
XM_011516695.2 1059 Intron XP_011514997.1
XM_011516696.2 1059 Intron XP_011514998.1
XM_011516697.2 1059 Intron XP_011514999.1
XM_017012794.1 1059 Intron XP_016868283.1
XM_017012795.1 1059 Intron XP_016868284.1
XM_017012796.1 1059 Intron XP_016868285.1
XM_017012797.1 1059 Intron XP_016868286.1
XM_017012798.1 1059 Intron XP_016868287.1
XM_017012799.1 1059 Intron XP_016868288.1
XM_017012800.1 1059 Intron XP_016868289.1
Gene
RNF133
Gene Name
ring finger protein 133
There are no transcripts associated with this gene.

Gene
RNF148
Gene Name
ring finger protein 148
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198085.1 1059 Missense Mutation CAT,CTT H281L NP_932351.1

View Full Product Details