Product Details
- SNP ID
-
rs150662833
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150801672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTCACCTGCTGCTCTGCGCTGC[A/G]GCCCCGGGCCACCCAGGCCAGGGGC
- Phenotype
-
MIM: 610334
MIM: 610385
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM176A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1127958] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM176A
- Gene Name
- transmembrane protein 176A
- Gene
- TMEM176B
- Gene Name
- transmembrane protein 176B
View Full Product Details