Product Details
- SNP ID
-
rs111773222
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:74603616 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTGCATTGCCAACTGGTGCAGC[A/T]TGGTCAAATCTGGCTGTGGAATGGC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR2052HG
PubMed Links
Gene Details
- Gene
- MIR2052HG
- Gene Name
- MIR2052 host gene
There are no transcripts associated with this gene.
View Full Product Details