Product Details

SNP ID
rs114145695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109243424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAAATGCCCAAGTAAAATATA[C/T]ACCATAAGGTATATAATTCTGAATC
Phenotype
MIM: 606109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDCD1 PubMed Links

Gene Details

Gene
NUDCD1
Gene Name
NudC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128211.1 Intron NP_001121683.1
NM_032869.3 Intron NP_116258.2
XM_017013910.1 Intron XP_016869399.1
XM_017013911.1 Intron XP_016869400.1

View Full Product Details