Product Details

SNP ID
rs114336567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6811311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCTCATGGGCAGTCCTCAGCT[C/G]TGGCAATGACACCCACACACCAACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XKR5 PubMed Links

Gene Details

Gene
XKR5
Gene Name
XK related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289973.1 2518 Missense Mutation CAG,GAG Q487E NP_001276902.1
NM_207411.4 2518 Missense Mutation CAG,GAG Q650E NP_997294.3
XM_011534750.2 2518 Missense Mutation CAG,GAG Q380E XP_011533052.1
XM_017013401.1 2518 Missense Mutation CAG,GAG Q652E XP_016868890.1
XM_017013402.1 2518 Missense Mutation CAG,GAG Q582E XP_016868891.1

View Full Product Details