Product Details

SNP ID
rs114645573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25462137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTGAAACAAACCATCTGATT[C/T]GTTTCATTGCTCGGCAGCAAAATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA2 PubMed Links

Gene Details

Gene
CDCA2
Gene Name
cell division cycle associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317906.1 859 Missense Mutation CGT,TGT R91C NP_001304835.1
NM_001317907.1 859 Missense Mutation CGT,TGT R106C NP_001304836.1
NM_152562.3 859 Missense Mutation CGT,TGT R106C NP_689775.2
XM_006716294.2 859 Missense Mutation CGT,TGT R111C XP_006716357.1
XM_011544415.1 859 Missense Mutation CGT,TGT R111C XP_011542717.1
XM_011544416.2 859 Missense Mutation CGT,TGT R111C XP_011542718.1
XM_011544417.2 859 Missense Mutation CGT,TGT R111C XP_011542719.1
XM_011544418.1 859 Missense Mutation CGT,TGT R106C XP_011542720.1
Gene
KCTD9
Gene Name
potassium channel tetramerization domain containing 9
There are no transcripts associated with this gene.

View Full Product Details