Product Details

SNP ID
rs114676196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80035771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGGATATTTTCTTCTACACTT[C/G]AGTCCTTACATCTGTGAGATTTCAT
Phenotype
MIM: 611990 MIM: 604068
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPS28 PubMed Links

Gene Details

Gene
MRPS28
Gene Name
mitochondrial ribosomal protein S28
There are no transcripts associated with this gene.

Gene
TPD52
Gene Name
tumor protein D52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025252.2 3116 UTR 3 NP_001020423.1
NM_001025253.2 3116 UTR 3 NP_001020424.1
NM_001287140.1 3116 UTR 3 NP_001274069.1
NM_001287142.1 3116 UTR 3 NP_001274071.1
NM_001287143.1 3116 UTR 3 NP_001274072.1
NM_001287144.1 3116 UTR 3 NP_001274073.1
NM_005079.3 3116 UTR 3 NP_005070.1

View Full Product Details