Product Details

SNP ID
rs116657732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:131908433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATAGGCTTAGTAGCAGTTTCCA[A/C]CTTACTGATTACTTTATGAAGACTT
Phenotype
MIM: 611798
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EFR3A PubMed Links

Gene Details

Gene
EFR3A
Gene Name
EFR3 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323553.1 Intron NP_001310482.1
NM_001323554.1 Intron NP_001310483.1
NM_001323555.1 Intron NP_001310484.1
NM_001323556.1 Intron NP_001310485.1
NM_001323557.1 Intron NP_001310486.1
NM_001323558.1 Intron NP_001310487.1
NM_015137.5 Intron NP_055952.2

View Full Product Details