Product Details

SNP ID
rs116759812
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:19404702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAAAGAGATTGTTTGGTTCACA[A/C]GGTATACTTTGGTTTATTTAAACAG
Phenotype
MIM: 616615 MIM: 614968
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CSGALNACT1 PubMed Links

Gene Details

Gene
CSGALNACT1
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130518.1 3991 UTR 3 NP_001123990.1
NM_018371.4 3991 UTR 3 NP_060841.5
XM_006716358.2 3991 UTR 3 XP_006716421.1
XM_006716359.2 3991 UTR 3 XP_006716422.1
XM_006716360.2 3991 UTR 3 XP_006716423.1
XM_006716361.2 3991 UTR 3 XP_006716424.1
XM_006716362.2 3991 UTR 3 XP_006716425.1
XM_006716363.1 3991 UTR 3 XP_006716426.1
XM_006716364.3 3991 UTR 3 XP_006716427.1
XM_011544578.1 3991 UTR 3 XP_011542880.1
XM_011544579.1 3991 UTR 3 XP_011542881.1
XM_011544580.1 3991 UTR 3 XP_011542882.1
XM_011544582.1 3991 UTR 3 XP_011542884.1
XM_011544583.1 3991 UTR 3 XP_011542885.1
XM_011544584.1 3991 UTR 3 XP_011542886.1
XM_011544585.2 3991 UTR 3 XP_011542887.1
XM_017013623.1 3991 UTR 3 XP_016869112.1
XM_017013624.1 3991 UTR 3 XP_016869113.1
XM_017013625.1 3991 UTR 3 XP_016869114.1
XM_017013626.1 3991 UTR 3 XP_016869115.1
XM_017013627.1 3991 UTR 3 XP_016869116.1
XM_017013628.1 3991 UTR 3 XP_016869117.1
XM_017013629.1 3991 UTR 3 XP_016869118.1
XM_017013630.1 3991 UTR 3 XP_016869119.1
XM_017013631.1 3991 UTR 3 XP_016869120.1
XM_017013632.1 3991 UTR 3 XP_016869121.1
XM_017013633.1 3991 UTR 3 XP_016869122.1
XM_017013634.1 3991 UTR 3 XP_016869123.1
XM_017013635.1 3991 UTR 3 XP_016869124.1
XM_017013636.1 3991 UTR 3 XP_016869125.1
XM_017013637.1 3991 UTR 3 XP_016869126.1
XM_017013638.1 3991 UTR 3 XP_016869127.1
XM_017013639.1 3991 UTR 3 XP_016869128.1
Gene
SH2D4A
Gene Name
SH2 domain containing 4A
There are no transcripts associated with this gene.

View Full Product Details