Product Details

SNP ID
rs117223605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11679922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTTTAATCCCTGTTGTGTGTC[G/T]CCGAATAGTTTACAAACTCTTTTTG
Phenotype
MIM: 600576
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GATA4 PubMed Links

Gene Details

Gene
GATA4
Gene Name
GATA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308093.1 Intron NP_001295022.1
NM_001308094.1 Intron NP_001295023.1
NM_002052.4 Intron NP_002043.2
XM_005272385.4 Intron XP_005272442.1
XM_005272386.1 Intron XP_005272443.1
XM_006716248.1 Intron XP_006716311.1
XM_011543817.2 Intron XP_011542119.1
XM_011543818.2 Intron XP_011542120.1
XM_017013312.1 Intron XP_016868801.1

View Full Product Details