Product Details

SNP ID
rs121918685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:127738437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAATTCGAGCTGCTGCCCACC[C/G]CGCCCCTGTCCCCTAGCCGCCGCTC
Phenotype
MIM: 190080
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CASC11 PubMed Links

Gene Details

Gene
CASC11
Gene Name
cancer susceptibility candidate 11 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MYC
Gene Name
v-myc avian myelocytomatosis viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002467.4 745 Missense Mutation CCG,GCG P74A NP_002458.2

View Full Product Details