Product Details

SNP ID
rs138379350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:87872873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCCCGAGGCGAGAAGAGAAGG[C/T]CACACTGGGAATGTCGTTCTCCGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF4L2 PubMed Links

Gene Details

Gene
DCAF4L2
Gene Name
DDB1 and CUL4 associated factor 4 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152418.3 1196 Missense Mutation NP_689631.1

View Full Product Details