Product Details

SNP ID
rs138825742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22138023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAGAATCAGGAGAAACAGAGCC[A/C]CTTTATGTATTTATTTATCAAAACA
Phenotype
MIM: 602302 MIM: 608302 MIM: 609349
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HR PubMed Links

Gene Details

Gene
HR
Gene Name
hair growth associated
There are no transcripts associated with this gene.

Gene
LGI3
Gene Name
leucine rich repeat LGI family member 3
There are no transcripts associated with this gene.

Gene
REEP4
Gene Name
receptor accessory protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316964.1 1552 UTR 3 NP_001303893.1
NM_001316965.1 1552 UTR 3 NP_001303894.1
NM_025232.3 1552 UTR 3 NP_079508.2

View Full Product Details