Product Details

SNP ID
rs139105121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81693672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAATTGTTAGATTAGGAAATTT[C/T]TTTTCCTTTTATCTTGTAAGAAAAA
Phenotype
MIM: 602064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPA1 PubMed Links

Gene Details

Gene
IMPA1
Gene Name
inositol monophosphatase 1
There are no transcripts associated with this gene.

Gene
SLC10A5
Gene Name
solute carrier family 10 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010893.2 1301 Missense Mutation NP_001010893.1
Gene
ZFAND1
Gene Name
zinc finger AN1-type containing 1
There are no transcripts associated with this gene.

View Full Product Details