Product Details

SNP ID
rs139490483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80631029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTTTTCTAAAGCTTTCTACCA[A/G]TGACCAAGGAGGTTCCCCGAACAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 12439 UTR 3 NP_001028895.1
XM_005251279.4 12439 UTR 3 XP_005251336.2
XM_017013725.1 12439 UTR 3 XP_016869214.1

View Full Product Details