Product Details

SNP ID
rs139955916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41929610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGCCGGCTGGCCCAGGAGCG[C/T]GTGACATGGAGAGATACAAAGGCAT
Phenotype
MIM: 601408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT6A PubMed Links

Gene Details

Gene
KAT6A
Gene Name
lysine acetyltransferase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305878.1 9266 Intron NP_001292807.1
NM_006766.4 9266 UTR 3 NP_006757.2
XM_011544656.2 9266 UTR 3 XP_011542958.1
XM_011544657.2 9266 UTR 3 XP_011542959.1
XM_011544658.2 9266 UTR 3 XP_011542960.1
XM_011544659.2 9266 UTR 3 XP_011542961.1
XM_017013863.1 9266 UTR 3 XP_016869352.1
XM_017013864.1 9266 UTR 3 XP_016869353.1

View Full Product Details