Product Details

SNP ID
rs140125530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125102341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACTAATTTCAAGATGCCAGGAC[A/G]TTCCAGTTCAAATTCAGGTTCAACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSMCE2 PubMed Links

Gene Details

Gene
NSMCE2
Gene Name
NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173685.2 277 Missense Mutation CAT,CGT H4R NP_775956.1
XM_005250875.2 277 Missense Mutation CAT,CGT H4R XP_005250932.1
XM_005250876.4 277 Missense Mutation CAT,CGT H4R XP_005250933.1
XM_011516974.2 277 Missense Mutation CAT,CGT H4R XP_011515276.1
XM_011516975.1 277 Missense Mutation CAT,CGT H4R XP_011515277.1
XM_017013330.1 277 Intron XP_016868819.1
XM_017013331.1 277 Missense Mutation CAT,CGT H4R XP_016868820.1
XM_017013332.1 277 Intron XP_016868821.1
XM_017013333.1 277 UTR 5 XP_016868822.1
XM_017013334.1 277 Intron XP_016868823.1

View Full Product Details