Product Details
- SNP ID
-
rs140513154
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:73292343 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCGAATTTCAGTTCTGTACATCTG[C/T]CTATATTCCTTGTGATAGTGCTTTG
- Phenotype
-
MIM: 607599
MIM: 604166
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C8orf89
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2070764] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C8orf89
- Gene Name
- chromosome 8 open reading frame 89
There are no transcripts associated with this gene.
- Gene
- RDH10
- Gene Name
- retinol dehydrogenase 10 (all-trans)
There are no transcripts associated with this gene.
- Gene
- RPL7
- Gene Name
- ribosomal protein L7
View Full Product Details