Product Details

SNP ID
rs140623927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125101493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCTATCTGATTCCATGAAAATA[C/T]CCCTTTTTCTGCTGCCATAGAGCCT
Phenotype
MIM: 610657
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0196 PubMed Links

Gene Details

Gene
KIAA0196
Gene Name
KIAA0196
There are no transcripts associated with this gene.

Gene
NSMCE2
Gene Name
NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173685.2 Intron NP_775956.1
XM_005250875.2 Intron XP_005250932.1
XM_005250876.4 Intron XP_005250933.1
XM_011516974.2 Intron XP_011515276.1
XM_011516975.1 Intron XP_011515277.1
XM_017013330.1 Intron XP_016868819.1
XM_017013331.1 Intron XP_016868820.1
XM_017013332.1 Intron XP_016868821.1
XM_017013333.1 Intron XP_016868822.1
XM_017013334.1 Intron XP_016868823.1

View Full Product Details