Product Details

SNP ID
rs141208943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24384518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACAACTTCATGCTGCGTGGGA[G/T]CTCCCAGCTACCTGCAGTGGCCACC
Phenotype
MIM: 606393
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMDEC1 PubMed Links

Gene Details

Gene
ADAMDEC1
Gene Name
ADAM like decysin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145271.1 234 UTR 5 NP_001138743.1
NM_001145272.1 234 UTR 5 NP_001138744.1
NM_014479.3 234 Missense Mutation AGC,ATC S5I NP_055294.1
Gene
LOC101929294
Gene Name
uncharacterized LOC101929294
There are no transcripts associated with this gene.

View Full Product Details