Product Details

SNP ID
rs142275114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143559442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGAGCTCCACTGGCTTCCAGC[A/C]CTACTGCCTGGTGGTTAGGAAGCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSDMD PubMed Links

Gene Details

Gene
GSDMD
Gene Name
gasdermin D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166237.1 810 Missense Mutation CAC,CCC H36P NP_001159709.1
NM_024736.6 810 Missense Mutation CAC,CCC H36P NP_079012.3
XM_011517301.1 810 Missense Mutation CAC,CCC H36P XP_011515603.1
Gene
MROH6
Gene Name
maestro heat like repeat family member 6
There are no transcripts associated with this gene.

Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details