Product Details

SNP ID
rs143752696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:12133965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGATGTGAGTTTTGAAAAGTG[A/G]TTGAACTGCCAGTCACCTCCCAAAT
Phenotype
MIM: 610186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM66D PubMed Links

Gene Details

Gene
FAM66D
Gene Name
family with sequence similarity 66 member D
There are no transcripts associated with this gene.

Gene
LOC392196
Gene Name
ubiquitin specific peptidase 17-like family member 2 pseudogene
There are no transcripts associated with this gene.

Gene
USP17L2
Gene Name
ubiquitin specific peptidase 17-like family member 2
There are no transcripts associated with this gene.

Gene
USP17L7
Gene Name
ubiquitin specific peptidase 17-like family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256869.1 Intron NP_001243798.1

View Full Product Details