Product Details

SNP ID
rs144867315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116770356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAAATCCTCATCATTATTTTG[C/T]GGCAACACAGGTGATACTACTTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP23 PubMed Links

Gene Details

Gene
UTP23
Gene Name
UTP23, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032334.2 454 Missense Mutation GCG,GTG A118V NP_115710.2

View Full Product Details