Product Details

SNP ID
rs145492318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143269401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGCAGGTCTCAGGGAAGATGG[C/T]AGCCCTAGGGGACATTCAGGAGTCC
Phenotype
MIM: 165280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLI4 PubMed Links

Gene Details

Gene
GLI4
Gene Name
GLI family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138465.3 90 Missense Mutation GCA,GTA A2V NP_612474.1
Gene
ZFP41
Gene Name
ZFP41 zinc finger protein
There are no transcripts associated with this gene.

View Full Product Details