Product Details

SNP ID
rs145986524
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86576067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTTCTTTTTGTTTATCTTCAT[C/T]TTCTTTTTGTTTATCTTCATTTTCT
Phenotype
MIM: 605080
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CNGB3 PubMed Links

Gene Details

Gene
CNGB3
Gene Name
cyclic nucleotide gated channel beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019098.4 2215 Missense Mutation AAT,GAT N723D NP_061971.3
XM_011517138.2 2215 Missense Mutation AAT,GAT N585D XP_011515440.1

View Full Product Details