Product Details

SNP ID
rs146829520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109243103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGAAATCCTAAAATAGGATCA[C/T]TGGTTTCTAGGCTTGCTACTTGCTG
Phenotype
MIM: 606109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDCD1 PubMed Links

Gene Details

Gene
NUDCD1
Gene Name
NudC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128211.1 1669 Missense Mutation AAT,AGT N524S NP_001121683.1
NM_032869.3 1669 Missense Mutation AAT,AGT N553S NP_116258.2
XM_017013910.1 1669 Missense Mutation AAT,AGT N466S XP_016869399.1
XM_017013911.1 1669 Missense Mutation AAT,AGT N466S XP_016869400.1

View Full Product Details