Product Details

SNP ID
rs147467210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:101501186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATGGGCATTCAGTGGAAAGCA[A/G]TCAATGCCTGTGGTAGATGAAGTAC
Phenotype
MIM: 608576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRHL2 PubMed Links

Gene Details

Gene
GRHL2
Gene Name
grainyhead like transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024915.3 Intron NP_079191.2
XM_011517305.2 Intron XP_011515607.1
XM_011517306.2 Intron XP_011515608.1
XM_011517307.2 Intron XP_011515609.1

View Full Product Details