Product Details

SNP ID
rs147680192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:70574953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTATGAAGATTTCTCTTTTTT[A/G]TTCCGGGGAGAGTCTGCTACATTTG
Phenotype
MIM: 605190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAM1 PubMed Links

Gene Details

Gene
TRAM1
Gene Name
translocation associated membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317804.1 1580 Silent Mutation AAC,AAT N337N NP_001304733.1
NM_001317805.1 1580 Silent Mutation AAC,AAT N282N NP_001304734.1
NM_014294.5 1580 Silent Mutation AAC,AAT N368N NP_055109.1
XM_017013266.1 1580 Silent Mutation AAC,AAT N272N XP_016868755.1
XM_017013267.1 1580 Silent Mutation AAC,AAT N272N XP_016868756.1

View Full Product Details