Product Details

SNP ID
rs147814759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41530208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCCTGGTTTCAGAGAAGATGAC[C/T]GAAGAAGTGGATTTCCTGGGACAGG
Phenotype
MIM: 610611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GINS4 PubMed Links

Gene Details

Gene
GINS4
Gene Name
GINS complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032336.2 125 Silent Mutation ACC,ACT T2T NP_115712.1
XM_005273659.3 125 Silent Mutation ACC,ACT T2T XP_005273716.1
Gene
LOC102723729
Gene Name
uncharacterized LOC102723729
There are no transcripts associated with this gene.

View Full Product Details