Product Details

SNP ID
rs148070033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:87872830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTCCTCCCGGACAGCCATGAGC[A/T]GCCCTGGTGCTCCTCGGAAGCCCCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF4L2 PubMed Links

Gene Details

Gene
DCAF4L2
Gene Name
DDB1 and CUL4 associated factor 4 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152418.3 1239 Missense Mutation CAG,CTG Q381L NP_689631.1

View Full Product Details