Product Details

SNP ID
rs148173181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123231323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGTTCGTATAAAAGAGGCACA[G/T]GCTTTCAGCTGAGTCTCTATCAACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf76 PubMed Links

Gene Details

Gene
C8orf76
Gene Name
chromosome 8 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032847.2 844 Silent Mutation GCA,GCC A264A NP_116236.1
Gene
ZHX1-C8orf76
Gene Name
ZHX1-C8orf76 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204180.1 844 Silent Mutation GCA,GCC A232A NP_001191109.1

View Full Product Details