Product Details

SNP ID
rs148347936
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39325241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACTTTATTAAGTTATTCCCTC[A/G]ATATATTGAAATGCATATTGTTGTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADAM5 PubMed Links

Gene Details

Gene
ADAM5
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details