Product Details

SNP ID
rs148753543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:115411181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATAAGATCTATCGTGTCAACG[C/T]TATCCAAAAATATTCTACTAAAAAG
Phenotype
MIM: 604386
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPS1 PubMed Links

Gene Details

Gene
TRPS1
Gene Name
transcriptional repressor GATA binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282902.2 7147 UTR 3 NP_001269831.1
NM_001282903.2 7147 UTR 3 NP_001269832.1
NM_014112.4 7147 UTR 3 NP_054831.2
XM_005251049.2 7147 UTR 3 XP_005251106.1
XM_006716625.1 7147 UTR 3 XP_006716688.1
XM_011517264.1 7147 UTR 3 XP_011515566.1
XM_011517266.2 7147 UTR 3 XP_011515568.1
XM_011517267.1 7147 UTR 3 XP_011515569.1
XM_011517268.1 7147 UTR 3 XP_011515570.1
XM_017013801.1 7147 UTR 3 XP_016869290.1

View Full Product Details