Product Details
- SNP ID
-
rs149778887
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144841581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAACTGTTTGAATGAGGAGACTGT[A/G]GTTCCCAAGACCTTCACCAAGGACG
- Phenotype
-
MIM: 608216
MIM: 194531
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COMMD5
PubMed Links
Gene Details
- Gene
- COMMD5
- Gene Name
- COMM domain containing 5
- Gene
- ZNF7
- Gene Name
- zinc finger protein 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282795.1 |
551 |
Silent Mutation |
GTA,GTG |
V169V |
NP_001269724.1 |
NM_001282796.1 |
551 |
Intron |
|
|
NP_001269725.1 |
NM_001282797.1 |
551 |
Silent Mutation |
GTA,GTG |
V62V |
NP_001269726.1 |
NM_003416.3 |
551 |
Silent Mutation |
GTA,GTG |
V158V |
NP_003407.1 |
XM_006716654.2 |
551 |
Silent Mutation |
GTA,GTG |
V62V |
XP_006716717.1 |
XM_006716656.3 |
551 |
Silent Mutation |
GTA,GTG |
V62V |
XP_006716719.1 |
XM_011517291.2 |
551 |
Silent Mutation |
GTA,GTG |
V175V |
XP_011515593.1 |
XM_011517292.2 |
551 |
Silent Mutation |
GTA,GTG |
V169V |
XP_011515594.1 |
XM_011517293.2 |
551 |
Silent Mutation |
GTA,GTG |
V158V |
XP_011515595.1 |
XM_011517294.2 |
551 |
Silent Mutation |
GTA,GTG |
V158V |
XP_011515596.1 |
XM_011517295.2 |
551 |
Silent Mutation |
GTA,GTG |
V157V |
XP_011515597.1 |
XM_011517296.2 |
551 |
Silent Mutation |
GTA,GTG |
V157V |
XP_011515598.1 |
XM_011517297.2 |
551 |
Silent Mutation |
GTA,GTG |
V62V |
XP_011515599.1 |
XM_017013813.1 |
551 |
Silent Mutation |
GTA,GTG |
V174V |
XP_016869302.1 |
XM_017013814.1 |
551 |
Silent Mutation |
GTA,GTG |
V179V |
XP_016869303.1 |
XM_017013815.1 |
551 |
Silent Mutation |
GTA,GTG |
V178V |
XP_016869304.1 |
XM_017013816.1 |
551 |
Intron |
|
|
XP_016869305.1 |
XM_017013817.1 |
551 |
Intron |
|
|
XP_016869306.1 |
View Full Product Details