Product Details

SNP ID
rs150307872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22281189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGTTTCTAAGACCCCTCTGAAA[C/T]GGGAAATGCTTCCATCAGGTATGTG
Phenotype
MIM: 610312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507071 PubMed Links

Gene Details

Gene
LOC100507071
Gene Name
uncharacterized LOC100507071
There are no transcripts associated with this gene.

Gene
PIWIL2
Gene Name
piwi like RNA-mediated gene silencing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135721.1 397 Missense Mutation CGG,TGG R90W NP_001129193.1
NM_018068.3 397 Missense Mutation CGG,TGG R90W NP_060538.2
XM_005273550.4 397 Missense Mutation CGG,TGG R90W XP_005273607.1
XM_005273551.3 397 Missense Mutation CGG,TGG R90W XP_005273608.1

View Full Product Details