Product Details

SNP ID
rs138448919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101592251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTGTACACTAATGTCCAGTTA[A/G]TTTTAGAGTGGGTGTAGCTAAGAAC
Phenotype
MIM: 606650 MIM: 613821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN3A PubMed Links

Gene Details

Gene
GRIN3A
Gene Name
glutamate ionotropic receptor NMDA type subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133445.2 2751 Intron NP_597702.2
XM_011518211.2 2751 Intron XP_011516513.1
Gene
PPP3R2
Gene Name
protein phosphatase 3 regulatory subunit B, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147180.3 2751 UTR 3 NP_671709.1

View Full Product Details