Product Details

SNP ID
rs138637921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100308513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATAATTACCTCTTTCTTAGAT[A/G]TTCTGCTTCCCCTTTCTCTATAGTG
Phenotype
MIM: 243305 MIM: 616717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INVS PubMed Links

Gene Details

Gene
INVS
Gene Name
inversin
There are no transcripts associated with this gene.

Gene
TEX10
Gene Name
testis expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161584.1 2156 Missense Mutation CAT,TAT H821Y NP_001155056.1
NM_017746.3 2156 Missense Mutation CAT,TAT H818Y NP_060216.2
XM_011518798.1 2156 Missense Mutation CAT,TAT H693Y XP_011517100.1
XM_017014848.1 2156 Missense Mutation CAT,TAT H463Y XP_016870337.1

View Full Product Details