Product Details

SNP ID
rs138966973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110201157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCTGGGCAGCCTCTGGGATTC[C/T]GGTTCCTTGCTGAGTCATCTGATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf152 PubMed Links

Gene Details

Gene
C9orf152
Gene Name
chromosome 9 open reading frame 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012993.2 1065 Missense Mutation AGA,GGA R171G NP_001013011.2

View Full Product Details