Product Details

SNP ID
rs139168222
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137048707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGCACCTGACGCAGCAGCAGGA[C/T]AAGCGCAGCCAGGAGCGCGGAGGCG
Phenotype
MIM: 602012 MIM: 605798
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ENTPD2 PubMed Links
Additional Information
For this assay, SNP(s) [rs17853460] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ENTPD2
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001246.3 1423 Missense Mutation ATC,GTC I457V NP_001237.1
NM_203468.2 1423 Missense Mutation ATC,GTC I480V NP_982293.1
XM_011519212.2 1423 Missense Mutation ATC,GTC I377V XP_011517514.1
Gene
NPDC1
Gene Name
neural proliferation, differentiation and control 1
There are no transcripts associated with this gene.

View Full Product Details